Assay Details
| Gene Symbol: ABHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016006.4 | NP_057090.2 | 4 - 5 | 766 | 81 | |
| XM_011533779.2 | XP_011532081.1 | 4 - 5 | 1016 | 81 | ||
| XM_011533780.1 | XP_011532082.1 | 4 - 5 | 713 | 81 | ||
| GenBank mRNA | AF151816.1 | - | 4 - 5 | 662 | 81 | |
| AK313811.1 | - | 4 - 5 | 676 | 81 | ||
| BC021958.1 | - | 4 - 5 | 694 | 3920855 | 81 | |
| KU178625.1 | - | 4 - 5 | 656 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000231.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
negative regulation of sequestering of triglyceride
positive regulation of triglyceride catabolic process
cell differentiation
positive regulation of lipoprotein lipase activity
1-acylglycerol-3-phosphate O-acyltransferase activity
triglyceride lipase activity
lysophosphatidic acid acyltransferase activity
carboxylic ester hydrolase activity
fatty acid metabolic process
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
negative regulation of sequestering of triglyceride
positive regulation of triglyceride catabolic process
cell differentiation
positive regulation of lipoprotein lipase activity
1-acylglycerol-3-phosphate O-acyltransferase activity
triglyceride lipase activity
lysophosphatidic acid acyltransferase activity
carboxylic ester hydrolase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03004681.hg.1 | TC03000231.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004681.hg.1 | ENST00000458276.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004681.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000013894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000454293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000456453.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000463153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | ENST00000486764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007194.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | ENST00000413300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | ENST00000458276.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | NM_016006.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150712.hg.1 | TC0300007194.hg.1 | Clariom D TC |

ABHD5
Genomic Map