Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005273018.1 | XP_005273075.1 | 1 - 2 | 190 | 73 | |
XM_017000106.1 | XP_016855595.1 | 1 - 2 | 190 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005273018.1, XM_017000106.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
