Assay Details
| Gene Symbol: ERO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019891.3 | NP_063944.3 | 13 - 14 | 1376 | 98 | |
| GenBank mRNA | AF252538.1 | - | 13 - 14 | 1293 | 98 | |
| AK315351.1 | - | 13 - 14 | 1295 | 98 | ||
| BC032823.2 | - | 13 - 14 | 1227 | 4829502 | 98 | |
| BC044573.1 | - | 13 - 14 | 1322 | 5270105 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01004004.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
4-hydroxyproline metabolic process
protein maturation by protein folding
insulin processing
extracellular matrix organization
glucose homeostasis
cell redox homeostasis
oxidation-reduction process
protein disulfide isomerase activity
protein binding
protein disulfide oxidoreductase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
unfolded protein binding
protein folding
4-hydroxyproline metabolic process
protein maturation by protein folding
insulin processing
extracellular matrix organization
glucose homeostasis
cell redox homeostasis
oxidation-reduction process
protein disulfide isomerase activity
protein binding
protein disulfide oxidoreductase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
unfolded protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01060614.hg.1 | TC01004004.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060614.hg.1 | ENST00000354619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01060614.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100234719.hg.1 | TC0100017849.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000264181.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000354619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | ENST00000366589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017849.hg.1 | NM_019891.3 | Refseq |

ERO1B
Genomic Map