Assay Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006045.2 | NP_006036.1 | 10 - 11 | 920 | 71 | |
| XM_011528481.2 | XP_011526783.1 | 10 - 11 | 948 | 71 | ||
| GenBank mRNA | AB014511.1 | - | 5 - 6 | 474 | 71 | |
| AK172802.1 | - | 8 - 9 | 609 | 71 | ||
| AK299064.1 | - | 10 - 11 | 849 | 71 | ||
| BC110591.1 | - | 6 - 7 | 412 | 71 | ||
| BC110592.1 | - | 6 - 7 | 412 | 71 | ||
| BF089115.1 | - | 2 - 3 | 185 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | TC2000009461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166174.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |

ATP9A
Genomic Map