Assay Details
| Gene Symbol: USP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199160.1 | NP_001186089.1 | 10 - 11 | 1789 | 93 | |
| NM_001199162.1 | NP_001186091.1 | 10 - 11 | 1759 | 93 | ||
| NM_006677.2 | NP_006668.1 | 9 - 10 | 1486 | 93 | ||
| XM_005264826.2 | XP_005264883.1 | 10 - 11 | 1790 | 93 | ||
| XM_006712948.2 | XP_006713011.1 | 10 - 11 | 1784 | 93 | ||
| XM_006712952.2 | XP_006713015.1 | 10 - 11 | 1739 | 93 | ||
| XM_006712953.1 | XP_006713016.1 | 10 - 11 | 1710 | 93 | ||
| XM_017005616.1 | XP_016861105.1 | 10 - 11 | 1781 | 93 | ||
| XM_017005618.1 | XP_016861107.1 | 10 - 11 | 1790 | 93 | ||
| XM_017005620.1 | XP_016861109.1 | 10 - 11 | 1781 | 93 | ||
| XM_017005623.1 | XP_016861112.1 | 10 - 11 | 1634 | 93 | ||
| XM_017005624.1 | XP_016861113.1 | 10 - 11 | 1634 | 93 | ||
| XM_017005626.1 | XP_016861115.1 | 9 - 10 | 1500 | 93 | ||
| XM_017005629.1 | XP_016861118.1 | 9 - 10 | 1557 | 93 | ||
| XM_017005632.1 | XP_016861121.1 | 9 - 10 | 1487 | 93 | ||
| XM_017005633.1 | XP_016861122.1 | 9 - 10 | 1481 | 93 | ||
| GenBank mRNA | AB020698.1 | - | 9 - 10 | 1328 | 93 | |
| AK294756.1 | - | 10 - 11 | 1682 | 93 | ||
| AK300425.1 | - | 10 - 11 | 1613 | 93 | ||
| BC106029.1 | - | 10 - 11 | 1607 | 93 | ||
| BC146752.1 | - | 9 - 10 | 1328 | 9007186 | 93 | |
More Information
Set Membership: |
|

USP19
Genomic Map