Assay Details
| Gene Symbol: DDOST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005216.4 | NP_005207.2 | 1 - 2 | 352 | 73 | |
| GenBank mRNA | AB062391.1 | - | 1 - 2 | 185 | 73 | |
| AK297009.1 | - | 1 - 2 | 279 | 73 | ||
| AK315633.1 | - | 1 - 2 | 332 | 73 | ||
| BC002594.2 | - | 1 - 2 | 259 | 3161588 | 73 | |
| BP233191.1 | - | 1 - 2 | 351 | 73 | ||
| CB988524.1 | - | 1 - 2 | 364 | 73 | ||
| D29643.1 | - | 1 - 2 | 315 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002316.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01035447.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035447.hg.1 | ENST00000375048.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035447.hg.1 | ENST00000415136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035447.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035447.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035450.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035450.hg.1 | ENST00000375048.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035450.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035446.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035446.hg.1 | ENST00000375048.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035446.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035446.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035451.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035451.hg.1 | ENST00000375048.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035451.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035452.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035452.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241659.hg.1 | TC0100018421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241653.hg.1 | TC0100018421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241660.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241660.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241660.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241660.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241660.hg.1 | TC0100018421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241654.hg.1 | TC0100018421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241658.hg.1 | TC0100018421.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 143257 | - | - |

DDOST
Genomic Map