Assay Details
| Gene Symbol: DDOST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005216.4 | NP_005207.2 | 2 - 3 | 458 | 128 | |
| GenBank mRNA | AB062391.1 | - | 2 - 3 | 291 | 128 | |
| AK297009.1 | - | 2 - 3 | 385 | 128 | ||
| AK315633.1 | - | 2 - 3 | 438 | 128 | ||
| BC002594.2 | - | 2 - 3 | 365 | 3161588 | 128 | |
| CB988524.1 | - | 2 - 3 | 470 | 128 | ||
| D29643.1 | - | 2 - 3 | 421 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002316.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01035444.hg.1 | TC01002316.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035444.hg.1 | ENST00000375048.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035444.hg.1 | ENST00000415136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01035444.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000464364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000475210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000477229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018421.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000375048.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000415136.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | ENST00000602624.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | NM_005216.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241651.hg.1 | TC0100018421.hg.1 | Clariom D TC |

DDOST
Genomic Map