Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009771.hg.1 |
TC06000820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009771.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009771.hg.1 |
ENST00000541107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009771.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009771.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161402.hg.1 |
TC0600008875.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000326771.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000450218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
ENST00000541107.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_001170807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008875.hg.1 |
NM_020482.5 |
Refseq |