Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007562.hg.1 |
ENST00000355086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007562.hg.1 |
ENST00000357825.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007562.hg.1 |
ENST00000543397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007562.hg.1 |
NM_020762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007562.hg.1 |
TC12000564.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000355086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000543397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000631006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
NM_020762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000355086.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000543397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
ENST00000631006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
NM_020762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158563.hg.1 |
TC1200007992.hg.1 |
Clariom D TC |