Assay Details
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024701.3 | NP_078977.2 | 3 - 4 | 426 | 84 | |
NR_037164.1 | 3 - 4 | 426 | 84 | |||
GenBank mRNA | AK023196.1 | - | 3 - 4 | 409 | 84 | |
AK292071.1 | - | 3 - 4 | 412 | 84 | ||
BC012056.1 | - | 3 - 4 | 403 | 4552643 | 84 | |
BP249999.1 | - | 3 - 4 | 426 | 84 | ||
CR457302.1 | - | 3 - 4 | 382 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001000.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | ENST00000357700.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012457.hg.1 | TC10001000.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10012454.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012454.hg.1 | TC10001000.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012460.hg.1 | ENST00000357700.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012460.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012460.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012460.hg.1 | TC10001000.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166092.hg.1 | TC1000009677.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166091.hg.1 | TC1000009677.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000357700.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000479033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000482921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | ENST00000493897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NM_024701.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_024581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009677.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166089.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166089.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166089.hg.1 | ENST00000459912.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166089.hg.1 | NR_037164.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166089.hg.1 | TC1000009677.hg.1 | Clariom D TC |
