Assay Details
| Gene Symbol: NAXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242883.1 | NP_001229812.1 | 1 - 2 | 188 | 145 | |
| GenBank mRNA | AK302117.1 | - | 1 - 2 | 73 | 145 | |
More Information
Set Membership: |
|

NAXD
Genomic Map