Assay Details
| Gene Symbol: ATP5C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001973.2 | NP_001001973.1 | 8 - 9 | 996 | 90 | |
| NM_001320886.1 | NP_001307815.1 | 7 - 8 | 961 | 90 | ||
| GenBank mRNA | BC000470.2 | - | 8 - 9 | 911 | 2961142 | 90 |
| BC000931.3 | - | 8 - 9 | 944 | 3445817 | 90 | |
| BC013394.2 | - | 8 - 9 | 911 | 2961142 | 90 | |
| BC013865.1 | - | 8 - 9 | 909 | 90 | ||
| BC016812.1 | - | 8 - 9 | 926 | 4092259 | 90 | |
| BC020824.1 | - | 8 - 9 | 932 | 4733791 | 90 | |
| BC026049.1 | - | 8 - 9 | 871 | 90 | ||
| D16562.1 | - | 8 - 9 | 923 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000067.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oxidative phosphorylation
ATP biosynthetic process
mitochondrial ATP synthesis coupled proton transport
ATPase activity
transmembrane transporter activity
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism
oxidative phosphorylation
ATP biosynthetic process
mitochondrial ATP synthesis coupled proton transport
ATPase activity
transmembrane transporter activity
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10000763.hg.1 | ENST00000356708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000763.hg.1 | ENST00000493053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000763.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000763.hg.1 | TC10000067.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000460820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000462760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000472202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006703.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000465936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145989.hg.1 | TC1000006703.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000335698.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000356708.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000473809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000480528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | ENST00000493053.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | NM_001001973.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | NM_005174.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000145986.hg.1 | TC1000006703.hg.1 | Clariom D TC |

ATP5C1
Genomic Map