Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034914.1 | NP_001030086.1 | 6 - 7 | 580 | 116 | |
| NM_013434.4 | NP_038462.1 | 7 - 8 | 773 | 116 | ||
| GenBank mRNA | AF120102.1 | - | 7 - 8 | 668 | 116 | |
| AF199599.1 | - | 6 - 7 | 658 | 116 | ||
| AF367022.1 | - | 6 - 7 | 592 | 116 | ||
| AJ131730.2 | - | 7 - 8 | 760 | 116 | ||
| AK315437.1 | - | 7 - 8 | 719 | 116 | ||
| BC012850.1 | - | 7 - 8 | 692 | 4179275 | 116 | |
| BT020075.1 | - | 6 - 7 | 658 | 116 | ||
| DQ148485.1 | - | 6 - 7 | 658 | 116 | ||
| DQ148486.1 | - | 6 - 7 | 580 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000572.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
intracellular protein transport
apoptotic process
signal transduction
cardiac conduction
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription corepressor activity
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
intracellular protein transport
apoptotic process
signal transduction
cardiac conduction
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription corepressor activity
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | TC02000572.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | ENST00000295225.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | ENST00000360990.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008766.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158624.hg.1 | TC0200008464.hg.1 | Clariom D TC |

KCNIP3
Genomic Map