Assay Details
| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001414.3 | NP_001405.1 | 5 - 6 | 704 | 59 | |
| GenBank mRNA | AL541292.3 | - | 5 - 6 | 672 | 59 | |
| BC103763.1 | - | 5 - 6 | 566 | 6148834 | 59 | |
| BC104189.2 | - | 4 - 5 | 495 | 40030991 | 59 | |
| CR456831.1 | - | 4 - 5 | 485 | 59 | ||
| X95648.1 | - | 4 - 5 | 495 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002092.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
response to heat
response to glucose
oligodendrocyte development
L-methionine biosynthetic process from methylthioadenosine
response to peptide hormone
positive regulation of GTPase activity
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity
translational initiation
regulation of translational initiation
response to heat
response to glucose
oligodendrocyte development
L-methionine biosynthetic process from methylthioadenosine
response to peptide hormone
positive regulation of GTPase activity
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | ENST00000228958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | ENST00000424014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | ENST00000539951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027619.hg.1 | TC12002092.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200196291.hg.1 | TC1200012281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000424014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000452159.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000534960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000537073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000539951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | ENST00000543940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012281.hg.1 | NM_001414.3 | Refseq |

EIF2B1
Genomic Map