| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
ENST00000282391.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
ENST00000380958.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000855.hg.1 |
TC13000085.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000363970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000856.hg.1 |
ENST00000282391.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000856.hg.1 |
ENST00000380958.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000856.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000856.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13000856.hg.1 |
TC13000085.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145753.hg.1 |
TC1300006715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300145747.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145747.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145747.hg.1 |
TC1300006715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300145750.hg.1 |
TC1300006715.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000363970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000380958.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000399613.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000483842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
ENST00000503791.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006715.hg.1 |
NM_175854.7 |
Refseq |