Assay Details
| Gene Symbol: HIST1H4H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003543.3 | NP_003534.1 | 1 | 61 | 78 | |
| GenBank mRNA | BC010926.1 | - | 1 | 100 | 78 | |
| BC120939.2 | - | 1 | 61 | 40115073 | 78 | |
| CR542169.1 | - | 1 | 61 | 78 | ||
| CR542187.1 | - | 1 | 61 | 78 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as Clariom D transcript cluster(s) TC0600011145.hg.1
					            	This assay targets the same location as Clariom D transcript cluster(s) TC0600011145.hg.1This assay targets the same location as HTA transcript cluster(s) TC06001355.hg.1
| Set Membership: |  Ambion siRNA available  Amplicon length between 71 and 85  Mammalian Gene Collection  Detect Genomic DNA  Homo sapiens  Both primers and probe map within a single exon | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 HIST1H4H
HIST1H4H Genomic Map
Genomic Map Back To Top
 Back To Top