Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005273013.4 | XP_005273070.1 | 5 - 6 | 675 | 82 | |
XM_005273018.1 | XP_005273075.1 | 5 - 6 | 682 | 82 | ||
XM_006711728.3 | XP_006711791.1 | 5 - 6 | 600 | 82 | ||
XM_011544028.2 | XP_011542330.1 | 5 - 6 | 600 | 82 | ||
XM_017000104.1 | XP_016855593.1 | 5 - 6 | 675 | 82 | ||
GenBank mRNA | AK297105.1 | - | 2 - 3 | 262 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000366541.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000435549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000463042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000476722.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000482234.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000490065.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000491888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000493334.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000496361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | ENST00000497459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012200.hg.1 | NM_006642.4 | Refseq |
