Assay Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133369.2 | NP_588610.2 | 12 - 13 | 2210 | 90 | |
XM_006714927.1 | XP_006714990.1 | 12 - 13 | 2190 | 90 | ||
XM_006714928.1 | XP_006714991.1 | 13 - 14 | 2358 | 90 | ||
XM_011534686.1 | XP_011532988.1 | 13 - 14 | 2358 | 90 | ||
GenBank mRNA | BC009333.2 | - | 6 - 7 | 1116 | 90 | |
BC122556.1 | - | 6 - 7 | 1087 | 90 | ||
BC157824.1 | - | 12 - 13 | 2093 | 9057043 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001007.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05014527.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014527.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014527.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014527.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167304.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167304.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167304.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167304.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167304.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
