Assay Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC033727.1 | - | 2 - 3 | 405 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC033727.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
