Assay Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006714928.1 | XP_006714991.1 | 6 - 7 | 1057 | 83 | |
XM_011534686.1 | XP_011532988.1 | 6 - 7 | 1057 | 83 | ||
XM_011534687.2 | XP_011532989.1 | 6 - 7 | 1057 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
