Assay Details
Gene Symbol: UNC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133369.2 | NP_588610.2 | 7 - 8 | 1265 | 60 | |
XM_006714927.1 | XP_006714990.1 | 7 - 8 | 1245 | 60 | ||
XM_006714928.1 | XP_006714991.1 | 8 - 9 | 1413 | 60 | ||
XM_011534686.1 | XP_011532988.1 | 8 - 9 | 1413 | 60 | ||
XM_011534687.2 | XP_011532989.1 | 8 - 9 | 1413 | 60 | ||
GenBank mRNA | BC009333.2 | - | 1 - 2 | 171 | 60 | |
BC033727.1 | - | 8 - 9 | 1393 | 60 | ||
BC122556.1 | - | 1 - 2 | 142 | 60 | ||
BC157824.1 | - | 7 - 8 | 1148 | 9057043 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001007.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05014520.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014520.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014520.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014520.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05014519.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014519.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014519.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014519.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05014522.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014522.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014522.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014522.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05014517.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014517.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014517.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014517.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05014521.hg.1 | TC05001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05014521.hg.1 | ENST00000261961.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014521.hg.1 | ENST00000329542.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05014521.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167295.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167298.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500167299.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167299.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167299.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167299.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167299.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167296.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500167297.hg.1 | TC0500009603.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
