Assay Details
| Gene Symbol: PCDHGB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018925.2 | NP_061748.1 | 1 - 2 | 2398 | 66 | |
| GenBank mRNA | AF152334.1 | - | 1 - 2 | 2398 | 66 | |
More Information
Set Membership: |
|

PCDHGB5
Genomic Map