Assay Details
| Gene Symbol: PCDHGB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018926.2 | NP_061749.1 | 1 - 2 | 2418 | 92 | |
| GenBank mRNA | AF152335.1 | - | 1 - 2 | 2418 | 92 | |
More Information
Set Membership: |
|

PCDHGB6
Genomic Map