Assay Details
Gene Symbol: PORCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282167.1 | NP_001269096.1 | 6 - 7 | 1018 | 67 | |
NM_022825.3 | NP_073736.2 | 6 - 7 | 873 | 67 | ||
XM_005272636.2 | XP_005272693.1 | 6 - 7 | 1228 | 67 | ||
XM_011543948.2 | XP_011542250.1 | 6 - 7 | 1157 | 67 | ||
XM_017029736.1 | XP_016885225.1 | 6 - 7 | 730 | 67 | ||
GenBank mRNA | AF317061.1 | - | 5 - 6 | 694 | 67 | |
AK056340.1 | - | 6 - 7 | 1200 | 67 | ||
AK057727.1 | - | 5 - 6 | 1120 | 67 | ||
AK293347.1 | - | 6 - 7 | 983 | 67 | ||
AK316378.1 | - | 5 - 6 | 765 | 67 | ||
AL579208.3 | - | 6 - 7 | 792 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
