Assay Details
Gene Symbol: TTC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193509.1 | NP_001180438.1 | 17 - 18 | 2226 | 68 | |
NM_017735.4 | NP_060205.3 | 17 - 18 | 2473 | 68 | ||
XM_005264416.1 | XP_005264473.1 | 16 - 17 | 2337 | 68 | ||
XM_011532958.1 | XP_011531260.1 | 16 - 17 | 2361 | 68 | ||
GenBank mRNA | AK000279.1 | - | 14 - 15 | 1681 | 68 | |
AK000665.1 | - | 12 - 13 | 1465 | 68 | ||
AK027570.1 | - | 17 - 18 | 2382 | 68 | ||
AK225295.1 | - | 14 - 15 | 1681 | 68 | ||
AK299196.1 | - | 17 - 18 | 2215 | 68 | ||
BC001248.1 | - | 16 - 17 | 2006 | 68 | ||
BC063791.1 | - | 17 - 18 | 2431 | 6500951 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000215.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02003193.hg.1 | TC02000215.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003193.hg.1 | ENST00000317907.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003193.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003193.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | ENST00000585007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007195.hg.1 | NR_039922.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200149688.hg.1 | TC0200007195.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000317907.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000428527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000433416.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000438654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000448773.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000454690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | ENST00000585007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_001193509.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NM_017735.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007195.hg.1 | NR_039922.1 | Refseq |
