Assay Details
| Gene Symbol: RNASEH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286834.1 | NP_001273763.1 | 4 - 5 | 624 | 100 | |
| NM_001286837.1 | NP_001273766.1 | 4 - 5 | 862 | 100 | ||
| NM_002936.4 | NP_002927.2 | 4 - 5 | 624 | 100 | ||
| GenBank mRNA | AF039652.1 | - | 4 - 5 | 594 | 100 | |
| AF048994.1 | - | 4 - 5 | 540 | 100 | ||
| AF048995.1 | - | 4 - 5 | 574 | 100 | ||
| AJ224117.1 | - | 4 - 5 | 513 | 100 | ||
| AK075490.1 | - | 4 - 5 | 573 | 100 | ||
| AK096913.1 | - | 4 - 5 | 780 | 100 | ||
| BC002973.1 | - | 4 - 5 | 583 | 3537074 | 100 | |
| BT019670.1 | - | 4 - 5 | 513 | 100 | ||
| CR541820.1 | - | 4 - 5 | 513 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004923.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02024845.hg.1 | TC02004923.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024845.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024845.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200225512.hg.1 | TC0200016618.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000315212.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000436842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000454734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | ENST00000464986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_001286837.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016618.hg.1 | NM_002936.5 | Refseq |

RNASEH1
Genomic Map