| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005672.hg.1 |
TC0X000426.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005672.hg.1 |
ENST00000373301.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005672.hg.1 |
ENST00000435339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005672.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152858.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152858.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152858.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152858.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152858.hg.1 |
TC0X00007713.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000514744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000607964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000610214.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007713.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
ENST00000435339.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
ENST00000614823.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
NM_001278000.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
NM_005296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152857.hg.1 |
TC0X00007713.hg.1 |
Clariom D TC |