Assay Details
| Gene Symbol: C8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015181.1 | - | 1 - 2 | 133 | 99 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000331434.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000524395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000529593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000530455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | ENST00000534350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009301.hg.1 | NM_023080.2 | Refseq |

C8orf33
Genomic Map