Assay Details
| Gene Symbol: RANBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006267.4 | NP_006258.3 | 23 - 24 | 8416 | 137 | |
| XM_005264003.2 | XP_005264060.1 | 24 - 25 | 8457 | 137 | ||
| GenBank mRNA | AB209483.1 | - | 20 - 21 | 8032 | 137 | |
| AK025711.1 | - | 4 - 5 | 2592 | 137 | ||
| AY170822.1 | - | 4 - 5 | 519 | 137 | ||
| D42063.1 | - | 23 - 24 | 8417 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000668.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY170822.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein peptidyl-prolyl isomerization
mRNA export from nucleus
tRNA export from nucleus
protein folding
NLS-bearing protein import into nucleus
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
negative regulation of glucokinase activity
intracellular transport of virus
regulation of gluconeogenesis involved in cellular glucose homeostasis
regulation of cellular response to heat
RNA binding
peptidyl-prolyl cis-trans isomerase activity
protein binding
zinc ion binding
Ran GTPase binding
ligase activity
SUMO transferase activity
protein peptidyl-prolyl isomerization
mRNA export from nucleus
tRNA export from nucleus
protein folding
NLS-bearing protein import into nucleus
sister chromatid cohesion
mitotic nuclear envelope disassembly
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
negative regulation of glucokinase activity
intracellular transport of virus
regulation of gluconeogenesis involved in cellular glucose homeostasis
regulation of cellular response to heat
RNA binding
peptidyl-prolyl cis-trans isomerase activity
protein binding
zinc ion binding
Ran GTPase binding
ligase activity
SUMO transferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02010475.hg.1 | TC02000668.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010475.hg.1 | ENST00000283195.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010475.hg.1 | ENST00000409491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010475.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000629728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | ENST00000629728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000425282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000495924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000629728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | ENST00000629728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008807.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200161536.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200161536.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200161536.hg.1 | ENST00000283195.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200161536.hg.1 | NM_006267.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200161536.hg.1 | TC0200008807.hg.1 | Clariom D TC |

RANBP2
Genomic Map