Assay Details
Gene Symbol: LY6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130478.1 | NP_001123950.1 | 3 - 4 | 286 | 118 | |
NM_001135655.1 | NP_001129127.1 | 2 - 3 | 266 | 118 | ||
NM_002347.4 | NP_002338.3 | 2 - 3 | 230 | 118 | ||
XM_017013413.1 | XP_016868902.1 | 1 - 2 | 244 | 118 | ||
XM_017013414.1 | XP_016868903.1 | 2 - 3 | 291 | 118 | ||
GenBank mRNA | AB012293.1 | - | 2 - 3 | 210 | 118 | |
AK314139.1 | - | 3 - 4 | 286 | 118 | ||
BC028894.1 | - | 2 - 3 | 256 | 4939118 | 118 | |
BC030192.1 | - | 2 - 3 | 190 | 5199071 | 118 | |
BI914261.1 | - | 1 - 2 | 71 | 117 | ||
BP347868.1 | - | 2 - 3 | 194 | 118 | ||
DB471677.1 | - | 2 - 3 | 230 | 118 | ||
DB502453.1 | - | 2 - 3 | 279 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001711.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08022516.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022516.hg.1 | ENST00000414417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022516.hg.1 | NM_001130478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022511.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022511.hg.1 | ENST00000342752.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022511.hg.1 | ENST00000414417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022511.hg.1 | NM_001130478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022511.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | ENST00000342752.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | ENST00000414417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | ENST00000430474.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | NM_001130478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022507.hg.1 | NM_002347.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022510.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022510.hg.1 | ENST00000430474.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022510.hg.1 | NM_002347.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022513.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022513.hg.1 | ENST00000342752.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022513.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | TC08001711.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | ENST00000342752.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | ENST00000414417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | ENST00000430474.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | NM_001130478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08022509.hg.1 | NM_002347.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182385.hg.1 | TC0800012110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012110.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182382.hg.1 | TC0800012110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182388.hg.1 | TC0800012110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182390.hg.1 | TC0800012110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000342752.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000414417.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000430474.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000479685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | ENST00000615409.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | NM_001135655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800182384.hg.1 | TC0800012110.hg.1 | Clariom D TC |
