Assay Details
| Gene Symbol: FUS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170634.1 | NP_001164105.1 | 3 - 4 | 293 | 110 | |
| XM_005255233.4 | XP_005255290.1 | 3 - 4 | 959 | 110 | ||
| GenBank mRNA | AK098774.1 | - | 3 - 4 | 264 | 110 | |
| X71428.1 | - | 3 - 4 | 260 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005255233.4 transcripts.
Set Membership: |
|

FUS
Genomic Map