Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004608.hg.1 |
ENST00000222249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004608.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004608.hg.1 |
TC19000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004607.hg.1 |
ENST00000222249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004607.hg.1 |
ENST00000536713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004607.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004607.hg.1 |
TC19000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004606.hg.1 |
ENST00000222249.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004606.hg.1 |
ENST00000536713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004606.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004606.hg.1 |
TC19000348.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000594192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000594192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000594192.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000594192.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000609922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007356.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
TC1900007356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153867.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
NM_002248.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
TC1900007356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
ENST00000222249.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
ENST00000615435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153866.hg.1 |
NM_002248.4 |
Refseq |