Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000132.3 | NP_000123.1 | 14 - 15 | 5388 | 69 | |
| GenBank mRNA | AX746360.1 | - | 14 - 15 | 5217 | 69 | |
| K01740.1 | - | 14 - 15 | 5367 | 69 | ||
| M14113.1 | - | 14 - 15 | 5388 | 69 | ||
| X01179.1 | - | 14 - 15 | 5326 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001547.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020749.hg.1 | TC0X001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020749.hg.1 | ENST00000360256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020749.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020751.hg.1 | TC0X001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020751.hg.1 | ENST00000360256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020751.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020748.hg.1 | TC0X001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020748.hg.1 | ENST00000360256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020748.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020750.hg.1 | TC0X001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020750.hg.1 | ENST00000360256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020750.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176805.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176805.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176805.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176805.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176805.hg.1 | TC0X00011226.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000330287.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000423959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000453950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | ENST00000483822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011226.hg.1 | NM_019863.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176806.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176806.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176806.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176806.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176806.hg.1 | TC0X00011226.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00176807.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176807.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176807.hg.1 | ENST00000360256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176807.hg.1 | NM_000132.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176807.hg.1 | TC0X00011226.hg.1 | Clariom D TC |

F8
Genomic Map