Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034077.hg.1 |
TC02002106.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034077.hg.1 |
ENST00000323853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034077.hg.1 |
ENST00000540328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034077.hg.1 |
ENST00000543553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034077.hg.1 |
NM_014014.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000323853.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000429650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000480615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
NM_014014.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000323853.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000429650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
ENST00000480615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
NM_014014.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198889.hg.1 |
TC0200013517.hg.1 |
Clariom D TC |