Assay Details
| Gene Symbol: MAP3K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006609.4 | NP_006600.3 | 1 - 2 | 105 | 113 | |
| GenBank mRNA | AK315007.1 | - | 2 - 3 | 289 | 113 | |
| BC136293.1 | - | 1 - 2 | 74 | 9020280 | 113 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002279.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC136293.1,NM_006609.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
activation of MAPK activity
protein phosphorylation
activation of JUN kinase activity
positive regulation of transcription, DNA-templated
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
metal ion binding
activation of MAPKK activity
activation of MAPK activity
protein phosphorylation
activation of JUN kinase activity
positive regulation of transcription, DNA-templated
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02036652.hg.1 | TC02002279.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036652.hg.1 | ENST00000344908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036652.hg.1 | ENST00000409947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036652.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036651.hg.1 | TC02002279.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036651.hg.1 | ENST00000344908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036651.hg.1 | ENST00000409947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036651.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036654.hg.1 | TC02002279.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036654.hg.1 | ENST00000344908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036654.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203511.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203511.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203511.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203511.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203511.hg.1 | TC0200014170.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203506.hg.1 | TC0200014170.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203509.hg.1 | TC0200014170.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 136013 | - | - |

MAP3K2
Genomic Map