Assay Details
| Gene Symbol: ABCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000033.3 | NP_000024.2 | 8 - 9 | 2268 | 82 | |
| GenBank mRNA | BC015541.1 | - | 8 - 9 | 2249 | 3896490 | 82 |
| BC025358.1 | - | 8 - 9 | 2268 | 4907640 | 82 | |
| Z21876.1 | - | 8 - 9 | 2255 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000755.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
peroxisome organization
peroxisomal long-chain fatty acid import
peroxisomal membrane transport
fatty acid beta-oxidation using acyl-CoA oxidase
alpha-linolenic acid metabolic process
long-chain fatty acid catabolic process
very long-chain fatty acid catabolic process
linoleic acid metabolic process
transmembrane transport
transporter activity
long-chain fatty acid transporter activity
peroxisomal fatty-acyl-CoA transporter activity
protein binding
ATP binding
ATPase activity
enzyme binding
ATPase activity, coupled to transmembrane movement of substances
identical protein binding
protein homodimerization activity
fatty acid beta-oxidation
peroxisome organization
peroxisomal long-chain fatty acid import
peroxisomal membrane transport
fatty acid beta-oxidation using acyl-CoA oxidase
alpha-linolenic acid metabolic process
long-chain fatty acid catabolic process
very long-chain fatty acid catabolic process
linoleic acid metabolic process
transmembrane transport
transporter activity
long-chain fatty acid transporter activity
peroxisomal fatty-acyl-CoA transporter activity
protein binding
ATP binding
ATPase activity
enzyme binding
ATPase activity, coupled to transmembrane movement of substances
identical protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009892.hg.1 | TC0X000755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009892.hg.1 | ENST00000218104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009892.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009893.hg.1 | TC0X000755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009893.hg.1 | ENST00000218104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009893.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010063.hg.1 | ENST00000562927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159935.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159935.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159935.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159935.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159935.hg.1 | TC0X00008795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007353.hg.1 | ENST00000429313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006472.hg.1 | ENST00000399687.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159934.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159934.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159934.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159934.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159934.hg.1 | TC0X00008795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008428.hg.1 | ENST00000445663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000218104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000370129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | ENST00000443684.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008795.hg.1 | NM_000033.3 | Refseq |

ABCD1
Genomic Map