Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006724828.3 | XP_006724891.1 | 1 - 2 | 1229 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006724828.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by vasopressin
positive regulation of systemic arterial blood pressure
renal water homeostasis
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
I-kappaB kinase/NF-kappaB signaling
excretion
hemostasis
positive regulation of cell proliferation
positive regulation of gene expression
telencephalon development
positive regulation of protein ubiquitination
interferon-gamma production
cellular response to hormone stimulus
response to cytokine
negative regulation of urine volume
negative regulation of renal sodium excretion
positive regulation of vasoconstriction
response to peptide
vasopressin receptor activity
protein binding
peptide binding
regulation of systemic arterial blood pressure by vasopressin
positive regulation of systemic arterial blood pressure
renal water homeostasis
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
I-kappaB kinase/NF-kappaB signaling
excretion
hemostasis
positive regulation of cell proliferation
positive regulation of gene expression
telencephalon development
positive regulation of protein ubiquitination
interferon-gamma production
cellular response to hormone stimulus
response to cytokine
negative regulation of urine volume
negative regulation of renal sodium excretion
positive regulation of vasoconstriction
response to peptide
vasopressin receptor activity
protein binding
peptide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000337474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000358927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000370049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000430697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | ENST00000434679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_000054.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NM_001146151.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008807.hg.1 | NR_027419.1 | Refseq |

AVPR2
Genomic Map