Assay Details
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104472.1 | 1 - 2 | 581 | 57 | ||
GenBank mRNA | AL543462.3 | - | 1 - 2 | 580 | 57 | |
AY040087.1 | - | 1 - 2 | 164 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
