Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000252457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000252458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000356221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000360383.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000375308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000375310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
ENST00000375312.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
NM_001078645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
NM_003903.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004929.hg.1 |
TC13000435.hg.1 |
HTA TC |