Assay Details
Gene Symbol: CDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006719993.1 | XP_006720056.1 | 1 - 2 | 234 | 88 | |
XM_017020832.1 | XP_016876321.1 | 1 - 2 | 234 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006719993.1, XM_017020832.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
