Assay Details
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021449.1 | - | 5 - 6 | 574 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
