Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002254.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002254.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002254.hg.1 |
TC20000188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002253.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002253.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002253.hg.1 |
TC20000188.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148230.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148230.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148230.hg.1 |
TC2000007073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148230.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148230.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148231.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148231.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148231.hg.1 |
TC2000007073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148231.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148231.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148232.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148232.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148232.hg.1 |
TC2000007073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148232.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148232.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
ENST00000201979.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007073.hg.1 |
NM_014012.5 |
Refseq |