Assay Details
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014252.3 | NP_055067.1 | 5 - 6 | 940 | 85 | |
GenBank mRNA | AF112968.1 | - | 5 - 6 | 777 | 85 | |
AF177333.1 | - | 5 - 6 | 742 | 85 | ||
AK296862.1 | - | 6 - 7 | 592 | 85 | ||
AK313602.1 | - | 5 - 6 | 822 | 85 | ||
BC002702.2 | - | 5 - 6 | 854 | 3614008 | 85 | |
BU726339.1 | - | 2 - 3 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000140.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300006628.hg.1 | ENST00000505530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006628.hg.1 | ENST00000505530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000384919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006960.hg.1 | NR_030352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | ENST00000338625.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | ENST00000379523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | ENST00000443985.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001630.hg.1 | TC13000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | ENST00000338625.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | ENST00000379523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | ENST00000443985.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001629.hg.1 | TC13000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13001631.hg.1 | ENST00000338625.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001631.hg.1 | ENST00000379523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001631.hg.1 | ENST00000443985.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001631.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001631.hg.1 | TC13000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | ENST00000338625.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | ENST00000379523.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | ENST00000443985.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001628.hg.1 | TC13000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007874.hg.1 | ENST00000508033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007874.hg.1 | ENST00000508033.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147207.hg.1 | TC1300006960.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300147208.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147208.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147208.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147208.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147208.hg.1 | TC1300006960.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300006628.hg.1 | ENST00000505530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006628.hg.1 | ENST00000505530.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147205.hg.1 | TC1300006960.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300147206.hg.1 | TC1300006960.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000338625.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000384919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000417731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000470509.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | ENST00000478827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NM_014252.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006960.hg.1 | NR_030352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006471.hg.1 | ENST00000457292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007564.hg.1 | ENST00000614260.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007874.hg.1 | ENST00000508033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007874.hg.1 | ENST00000508033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00007282.hg.1 | ENST00000456738.1 | Ensembl |
