Assay Details
Gene Symbol: ELF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005266277.3 | XP_005266334.1 | 1 - 2 | 573 | 112 | |
XM_011534951.2 | XP_011533253.1 | 1 - 2 | 573 | 112 | ||
GenBank mRNA | BQ722965.1 | - | 1 - 2 | 38 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005266277.3, XM_011534951.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytokine production
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of T cell receptor signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
regulation of cytokine production
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of T cell receptor signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
