Assay Details
Gene Symbol: ELF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172373.3 | NP_758961.1 | 4 - 5 | 726 | 77 | |
XM_005266276.1 | XP_005266333.1 | 4 - 5 | 615 | 77 | ||
XM_005266277.3 | XP_005266334.1 | 4 - 5 | 1158 | 77 | ||
XM_011534950.1 | XP_011533252.1 | 6 - 7 | 964 | 77 | ||
XM_011534951.2 | XP_011533253.1 | 5 - 6 | 1277 | 77 | ||
XM_017020409.1 | XP_016875898.1 | 6 - 7 | 944 | 77 | ||
XM_017020410.1 | XP_016875899.1 | 5 - 6 | 881 | 77 | ||
XM_017020411.1 | XP_016875900.1 | 5 - 6 | 827 | 77 | ||
XM_017020412.1 | XP_016875901.1 | 5 - 6 | 955 | 77 | ||
XM_017020413.1 | XP_016875902.1 | 5 - 6 | 1009 | 77 | ||
GenBank mRNA | BC030507.2 | - | 3 - 4 | 581 | 4385989 | 77 |
M82882.1 | - | 4 - 5 | 668 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000597.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytokine production
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of T cell receptor signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
regulation of cytokine production
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of T cell receptor signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13006827.hg.1 | ENST00000239882.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006827.hg.1 | ENST00000498824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13006827.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13006827.hg.1 | TC13000597.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000405737.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000442101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008702.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | ENST00000625359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | NM_001145353.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158777.hg.1 | TC1300008702.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | ENST00000239882.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | ENST00000498824.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | NM_172373.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300158778.hg.1 | TC1300008702.hg.1 | Clariom D TC |
