Assay Details
| Gene Symbol: RPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002950.3 | NP_002941.1 | 3 - 4 | 705 | 81 | |
| GenBank mRNA | AK223595.1 | - | 3 - 4 | 678 | 81 | |
| AK296905.1 | - | 3 - 4 | 581 | 81 | ||
| AK297513.1 | - | 3 - 4 | 690 | 81 | ||
| AK297941.1 | - | 4 - 5 | 574 | 81 | ||
| AK312369.1 | - | 3 - 4 | 647 | 81 | ||
| BC007995.1 | - | 3 - 4 | 512 | 81 | ||
| BC010839.1 | - | 3 - 4 | 666 | 3461167 | 81 | |
| CR749284.1 | - | 3 - 4 | 659 | 81 | ||
| Y00281.1 | - | 3 - 4 | 766 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001757.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297941.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | TC03001757.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | ENST00000296255.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | ENST00000497289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | ENST00000537139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | ENST00000545956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031875.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | TC03001757.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | ENST00000296255.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | ENST00000497289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | ENST00000537139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | ENST00000545956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031873.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | TC03001757.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | ENST00000296255.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | ENST00000497289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | ENST00000537139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | ENST00000545956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031872.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031876.hg.1 | TC03001757.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031876.hg.1 | ENST00000296255.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031876.hg.1 | ENST00000497289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031876.hg.1 | ENST00000545956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031876.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192364.hg.1 | TC0300012346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000476931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000479113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000481168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000490166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000495462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | ENST00000497415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012346.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | ENST00000296255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | ENST00000497289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | NM_002950.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192363.hg.1 | TC0300012346.hg.1 | Clariom D TC |

RPN1
Genomic Map