Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029133.hg.1 |
TC03001612.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029133.hg.1 |
ENST00000284322.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029133.hg.1 |
ENST00000471714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029133.hg.1 |
ENST00000495063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029133.hg.1 |
NM_015429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000284322.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000459682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000471714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000483129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000495063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000528305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000533855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
NM_015429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000284322.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000459682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000471714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000483129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000495063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000528305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
ENST00000533855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
NM_015429.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188074.hg.1 |
TC0300011849.hg.1 |
Clariom D TC |