Assay Details
| Gene Symbol: HERC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165136.1 | NP_001158608.1 | 11 - 12 | 1547 | 83 | |
| NM_017912.3 | NP_060382.3 | 11 - 12 | 1547 | 83 | ||
| XM_005263083.3 | XP_005263140.1 | 11 - 12 | 1744 | 83 | ||
| XM_011532053.2 | XP_011530355.1 | 11 - 12 | 1746 | 83 | ||
| XM_017008333.1 | XP_016863822.1 | 10 - 11 | 1105 | 83 | ||
| GenBank mRNA | AF336798.1 | - | 11 - 12 | 1547 | 83 | |
| AK097168.1 | - | 5 - 6 | 362 | 83 | ||
| AK295832.1 | - | 11 - 12 | 1457 | 83 | ||
| AY653201.1 | - | 11 - 12 | 1547 | 83 | ||
| AY653202.1 | - | 10 - 11 | 1425 | 83 | ||
| AY653203.1 | - | 9 - 10 | 1288 | 83 | ||
| BC042047.1 | - | 11 - 12 | 1423 | 5531519 | 83 | |
| BX647121.1 | - | 11 - 12 | 1223 | 83 | ||
More Information
Set Membership: |
|

HERC6
Genomic Map