Assay Details
| Gene Symbol: BCL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282237.1 | NP_001269166.1 | 3 - 4 | 1127 | 73 | |
| NM_138576.3 | NP_612808.1 | 3 - 4 | 1130 | 73 | ||
| GenBank mRNA | AB041218.1 | - | 3 - 4 | 1130 | 73 | |
| AB043584.1 | - | 3 - 4 | 645 | 73 | ||
| AB057668.1 | - | 3 - 4 | 1127 | 73 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
keratinocyte development
epithelial cell morphogenesis
transcription from RNA polymerase II promoter
signal transduction
negative regulation of cell proliferation
regulation of keratinocyte proliferation
regulation of lipid metabolic process
striatal medium spiny neuron differentiation
commitment of neuronal cell to specific neuron type in forebrain
neurogenesis
post-embryonic camera-type eye development
T cell differentiation in thymus
T cell receptor V(D)J recombination
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
positive T cell selection
regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
alpha-beta T cell differentiation
thymus development
olfactory bulb axon guidance
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
keratinocyte development
epithelial cell morphogenesis
transcription from RNA polymerase II promoter
signal transduction
negative regulation of cell proliferation
regulation of keratinocyte proliferation
regulation of lipid metabolic process
striatal medium spiny neuron differentiation
commitment of neuronal cell to specific neuron type in forebrain
neurogenesis
post-embryonic camera-type eye development
T cell differentiation in thymus
T cell receptor V(D)J recombination
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
positive T cell selection
regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
alpha-beta T cell differentiation
thymus development
olfactory bulb axon guidance
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000345514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000357195.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | ENST00000443726.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_001282238.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_022898.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010156.hg.1 | NM_138576.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 144845 | - | - |

BCL11B
Genomic Map