Assay Details
Gene Symbol: MED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052877.4 | NP_443109.2 | 1 - 2 | 56 | 84 | |
NM_201542.4 | NP_963836.2 | 1 - 2 | 56 | 84 | ||
GenBank mRNA | BG620658.1 | - | 1 - 2 | 57 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002584.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_052877.4,NM_201542.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | TC01002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | ENST00000290663.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | ENST00000372457.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040208.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | TC01002584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | ENST00000290663.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | ENST00000372457.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040210.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100204164.hg.1 | TC0100013925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
